Silver ≥99.999%, powder

Supplier: STREM CHEMICALS INC MS

93-4757-5G 93-4757-25G
100207-082EA 108.98 USD
100207-082 100200-150
Silver ≥99.999%, powder
Silver
Formula: Ag
Boiling Pt: 2212 °C
Melting Pt: 960.5 °C
Density: 10.49
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR