Silver chromate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Argentous chromate, Silver(I) chromate

93-4745-50G 93-4745-10G
100208-184EA 390.65 USD
100208-184 100202-146
Silver chromate ≥99%
Silver chromate
Formula: Ag₂CrO₄
Boiling Pt: 1550 °C (1013 hPa)
Density: 5.625
MDL Number: MFCD00003402
CAS Number: 7784-01-2
EINECS: 232-043-8
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR