Silver iodide ≥99.9% (Ag basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Argentous iodide, Silver monoiodide
93-4714-10G
93-4714-50G
100204-334EA
94.83
USD
100204-334
100203-476
Silver iodide ≥99.9% (Ag basis)
Silver iodide
Formula:
AgI Boiling Pt: 1506 °C Melting Pt: 558 °C Density: 6.010 |
MDL Number:
MFCD00003412 CAS Number: 7783-96-2 EINECS: 232-038-0 Merck Index: 12,08659 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...