Palladium ≥99.999%, powder, PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-4630-1G
93-4630-5G
100201-114EA
351.02
USD
100201-114
100203-474
Palladium ≥99.999%, powder, PURATREM
Palladium
Formula:
Pd Boiling Pt: 3140 °C Melting Pt: 1552 °C Density: 12.02 Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...