Niobium ≥99.8%, turnings
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4130-100G
93-4130-25G
100204-724EA
125.97
USD
100204-724
100205-572
Niobium ≥99.8%, turnings
Niobium
Formula:
Nb MW: 92.91 g/mol Boiling Pt: 3700 Melting Pt: 2468 Density: 8.57 |
MDL Number:
MFCD00011126 CAS Number: 7440-03-1 EINECS: 231-113-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...