Niobium(V) chloride ≥99% (Nb basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Niobium pentachloride
93-4126-250G
93-4126-50G
100208-174EA
339.69
USD
100208-174
100203-470
Niobium(V) chloride ≥99% (Nb basis)
Niobium (V) chloride
Formula:
Cl₅Nb Boiling Pt: 254 °C Melting Pt: 204…206 °C Density: 2.75 |
MDL Number:
MFCD00011127 CAS Number: 10026-12-7 EINECS: 233-059-8 UN: 3260 ADR: 8,II Merck Index: 12,06649 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...