Zirconium(IV) oxide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Zirconium dioxide, Zirconium Oxide

93-4013-250G 93-4013-1000G
100210-396EA 65.11 USD
100210-396 100203-854
Zirconium(IV) oxide ≥99%
Zirconium (IV) oxide
Formula: ZrO₂
MW: 123.22 g/mol
Boiling Pt: 4300
Melting Pt: 2680
Density: 5.6
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR