Yttrium(III) acetylacetonate hydrate ≥99.9% (Y basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-3901-25G
93-3901-5G
93-3901-100G
100197-838EA
67.94
USD
100197-838
100198-372
100208-718
Yttrium(III) acetylacetonate hydrate ≥99.9% (Y basis)
Yttrium (III) acetylacetonate hydrate
Formula:
C₁₅H₂₁O₆Y·H₂O MW: 404.25 g/mol |
MDL Number:
MFCD00013509 CAS Number: 207801-29-4 EINECS: 239-607-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...