Rubidium ≥99%

Supplier: STREM CHEMICALS INC MS
Danger


Prescored ampoule.

93-3735-1G 93-3735-5G
100198-370EA 227.88 USD
100198-370 100204-714
Rubidium ≥99%
Rubidium
Formula: Rb
Boiling Pt: 688 °C
Melting Pt: 38.9 °C
Density: 1.532
MDL Number: MFCD00134055
CAS Number: 7440-17-7
EINECS: 231-126-6
UN: 1423
ADR: 4.3,I
Merck Index: 12,08439

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR