Bromine ≥99.5%
Supplier: 0000018203
|
Danger
|
93-3501-250G
93-3501-1000G
100210-876EA
195.32
USD
100210-876
100205-540
Bromine ≥99.5%
Bromine
Formula:
Br₂ MW: 159.81 g/mol Boiling Pt: 58.8 °C Melting Pt: –7.2 °C Density: 3.12 Storage Temperature: Ambient |
MDL Number:
MFCD00010896 CAS Number: 7726-95-6 EINECS: 231-778-1 UN: 1744 ADR: 8,I Merck Index: 13,01378 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...