Arsenic (V) oxide ≥99.9% (As basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Arsenic oxide, Arsenic pentoxide, di-Arsenic pentaoxide

93-3304-50G 93-3304-10G
100207-294EA 529.35 USD
100207-294 100199-074
Arsenic (V) oxide ≥99.9% (As basis)
Arsenic (V) oxide
Formula: As₂O₅
MW: 229.84 g/mol
Melting Pt: 730 (dec.)
Density: 4.32
MDL Number: MFCD00003434
CAS Number: 1303-28-2
EINECS: 215-116-9
UN: 1559
ADR: 6.1 II
Merck Index: 12,00839

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR