Germanium(IV) ethoxide ≥99.99% (Ge basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Germanium tetraethanolate, Germanic acid tetraethyl ester, Tetraethyl orthogermanate, Germanium tetraethoxide, Germanium (IV) ethanolate

93-3206-25G 93-3206-1G 93-3206-5G
100207-034EA 860.55 USD
100207-034 100210-362 100207-036
Germanium(IV) ethoxide ≥99.99% (Ge basis), PURATREM
Germanium (IV) ethoxide

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formula: C₈H₂₀GeO₄
MW: 252.85 g/mol
Boiling Pt: 185.5 °C
Melting Pt: -72
Density: 1.14
MDL Number: MFCD00015121
CAS Number: 14165-55-0
EINECS: 238-007-8
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR