Zinc sulfate monohydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-3045-250G 93-3045-1000G
100197-818EA 36.8 USD
100197-818 100202-078
Zinc sulfate monohydrate ≥99%
Zinc sulphate monohydrate
Formula: ZnSO₄·1H₂O
MW: 179.45 g/mol
Density: 3.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149893
CAS Number: 7446-19-7
EINECS: 231-793-3
UN: 3077
ADR: 9 III
Merck Index: 13,10213

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR