Zinc selenide ≥99.99% (Zn basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Zinc (II) selenide
93-3041-100G
93-3041-25G
93-3041-5G
100203-444EA
861.97
USD
100203-444
100204-308
100210-352
Zinc selenide ≥99.99% (Zn basis), PURATREM
Zinc selenide
Formula:
SeZn Melting Pt: > 1100 °C Density: 5.42 Storage Temperature: Ambient |
MDL Number:
MFCD00011296 CAS Number: 1315-09-9 EINECS: 215-259-7 UN: 3283 ADR: 6.1,III Merck Index: 12,10290 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...