Zinc sulfide ≥99.9% (Zn basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Zinc (II) sulphide, Zinc (II) sulfide

93-3025-25G 93-3025-100G
100204-302EA 50.95 USD
100204-302 100209-092
Zinc sulfide ≥99.9% (Zn basis)
Zinc sulphide
Formula: SZn
MW: 97.46 g/mol
Melting Pt: 1064
Density: 4.10
Storage Temperature: Ambient
MDL Number: MFCD00011301
CAS Number: 1314-98-3
EINECS: 215-251-3
Merck Index: 12,10294

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR