Scandium ≥99.9%, powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-2120-2G
93-2120-500MG
100206-962EA
1341.78
USD
100206-962
100199-004
Scandium ≥99.9%, powder
Scandium
Formula:
Sc MW: 44.96 g/mol Boiling Pt: 2730 Melting Pt: 1541 Density: 3.0 |
MDL Number:
MFCD00016323 CAS Number: 7440-20-2 EINECS: 231-129-2 UN: 3089 ADR: 4.1 III Merck Index: 12,08537 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...