Platinum ≥99.99%, foil

Supplier: STREM CHEMICALS INC MS

78-0005-25X25MM 78-0005-50X50MM
100203-302EA 326.95 USD
100203-302 100203-300
Platinum ≥99.99%, foil
Platinum
Formula: Pt
Boiling Pt: 3827 °C
Melting Pt: 1769 °C
Density: 21.45
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR