2-Amino-2-methyl-1,3-propanediol 97 for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

A1117-25GM A1117-500GM A1117-100GM
700006-968EA 92.25 USD
700006-968 700006-970 700006-966
2-Amino-2-methyl-1,3-propanediol 97 for synthesis
2-Amino-2-methyl-1,3-propanediol

2-Amino-2-methyl-1,3-propanediol is also called AMPD and often used as a reaction buffer for enzymes with an activity in the basic pH range.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 272
Melting Pt: 102…105 °C
Density: 1.211 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004678
CAS Number: 115-69-5
EINECS: 204-100-7
Merck Index: 13,00447

Order Now

Specification Test Results

Assay 97% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR