Copper(II) chloride dihydrate ≥99.999% (Cu basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

93-2985-50G 93-2985-10G
100202-066EA 386.4 USD
100202-066 100204-298
Copper(II) chloride dihydrate ≥99.999% (Cu basis), PURATREM
Copper (II) chloride dihydrate
Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100
Density: 2.54
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8 III
Merck Index: 13,02660

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR