Copper ≥99.999%, powder, very pure

Supplier: STREM CHEMICALS INC MS
Danger

93-2971-25G 93-2971-5G
100196-606EA 461.42 USD
100196-606 100196-608
Copper ≥99.999%, powder, very pure
Copper
Formula: Cu
Boiling Pt: 2567 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.92
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR