Copper(II) acetylacetonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate

93-2968-50G 93-2968-250G
100196-604EA 67.94 USD
100196-604 100198-354
Copper(II) acetylacetonate ≥98%
Copper (II) acetylacetonate
Formula: C₁₀H₁₄CuO₄
Boiling Pt: 284 °C/ 78 °C (0.05 mmHg) (dec./ subl.)
Melting Pt: 284…285 °C (dec.)
MDL Number: MFCD00000016
CAS Number: 13395-16-9
EINECS: 236-477-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR