Copper(II) sulfate pentahydrate ≥99.999% (Cu basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-2959-100G
93-2959-25G
93-2959-5G
100203-850EA
547.75
USD
100203-850
100209-084
100202-056
Copper(II) sulfate pentahydrate ≥99.999% (Cu basis), PURATREM
Copper (II) sulphate pentahydrate
Formula:
CuSO₄·5H₂O Melting Pt: 110 °C (dec.) Density: 2.284 Storage Temperature: Ambient |
MDL Number:
MFCD00149681 CAS Number: 7758-99-8 EINECS: 231-847-6 UN: 3077 ADR: 9,III REACH: 01-2119520566-40 Merck Index: 13,02682 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...