Copper(II) sulfate pentahydrate ≥98% ACS

Supplier: STREM CHEMICALS INC MS
Warning

93-2958-1000G 93-2958-250G
100205-496EA 176.92 USD
100205-496 100196-602
Copper(II) sulfate pentahydrate ≥98% ACS
Copper (II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
Melting Pt: 110 °C (dec.)
Density: 2.284
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR