Nickel ≥99%, wire
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-2869-10MT
93-2869-50MT
100206-998EA
110.4
USD
100206-998
100200-110
Nickel ≥99%, wire
Nickel
Formula:
Ni Boiling Pt: 2732 °C Melting Pt: 1453 °C Density: 8.90 Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,III Merck Index: 12,06582 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...