Ammonium iron (II) sulfate hexahydrate ≥98.5% ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Ferrous Ammonium sulphate Hexahydrate
, Ferrous ammonium sulfate hexahydrate
, Ammonium iron(Ⅱ) sulfate hexahydrate
, Ammonium ferrous sulphate hexahydrate
, Ferrous Ammonium sulfate Hexahydrate
, di-Ammonium iron (II) sulphate hexahydrate
, Ammonium ferrous sulfate hexahydrate
, Ferrous ammonium sulphate hexahydrate
, Ammonium iron(II) sulphate hydrate
, di-Ammonium iron (II) sulfate hexahydrate
, Ammonium iron(Ⅱ) sulphate hexahydrate
, Ammonium iron(II) sulfate hydrate
93-2650-2000G
93-2650-500G
100199-030EA
474.15
USD
100199-030
100210-300
Ammonium iron (II) sulfate hexahydrate ≥98.5% ACS
Ammonium iron (II) sulphate hexahydrate
Formula:
(NH₄)₂Fe(SO₄)₂·6H₂O MW: 392.14 g/mol Melting Pt: 100…110 °C (dec.) Density: 1.864 Storage Temperature: Ambient |
MDL Number:
MFCD00150530 CAS Number: 7783-85-9 EINECS: 233-151-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...