Manganese (II) acetylacetonate ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')manganese (II), Manganese (II) bis(2,4-pentanedionato-O,O'), Manganese(II) bis(acetylacetonate), Manganese (II) 2,4-pentanedionate

93-2503-250G 93-2503-50G
100202-010EA 587.38 USD
100202-010 100204-246
Manganese (II) acetylacetonate ≥95%
Manganese (II) acetylacetonate
Formula: C₁₀H₁₄MnO₄
MW: 253.16 g/mol
Melting Pt: > 180 °C
MDL Number: MFCD00000022
CAS Number: 14024-58-9
EINECS: 237-858-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR