Manganese(II) acetate tetrahydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-2502-250G 93-2502-1000G
100199-022EA 45.29 USD
100199-022 100197-766
Manganese(II) acetate tetrahydrate ≥99%
Manganese (II) acetate tetrahydrate
Formula: (H₃CCOO)₂Mn·4H₂O
Melting Pt: 300 °C
Density: 1.589
MDL Number: MFCD00062552
CAS Number: 6156-78-1
EINECS: 211-334-3
Merck Index: 13,05746

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR