Chromium ≥99.3%, flakes
Supplier: STREM CHEMICALS INC MS
93-2456-500G
93-2456-100G
100210-278EA
124.55
USD
100210-278
100202-006
Chromium ≥99.3%, flakes
Chromium
Formula:
Cr Boiling Pt: 2200 °C Melting Pt: 1890 °C Density: 7.20 Storage Temperature: Ambient |
MDL Number:
MFCD00010944 CAS Number: 7440-47-3 EINECS: 231-157-5 Merck Index: 12,02288 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...