Vanadium ≥99.5%, powder

Supplier: STREM CHEMICALS INC MS

93-2341-25G 93-2341-5G
100201-266EA 268.92 USD
100201-266 100200-538
Vanadium ≥99.5%, powder
Vanadium
Formula: V
MW: 50.94 g/mol
Boiling Pt: 3477
Melting Pt: 1896
Density: 6.11
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
UN: 3089
ADR: 4.1 II
Merck Index: 12,10053

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR