Titanium ≥99.7%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-2260-500G
93-2260-25G
93-2260-100G
100201-988EA
336.86
USD
100201-988
100197-752
100206-970
Titanium ≥99.7%, powder
Titanium
Formula:
Ti MW: 47.88 g/mol Boiling Pt: 3262 Melting Pt: 1660 Density: 4.5 |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 UN: 2546 ADR: 4.2 II Merck Index: 12,09610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...