Titanium(IV) isopropoxide ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Titanium (IV) isopropoxideTitanium tetraisopropanolate

93-2216-1000G 93-2216-250G
100199-514EA 225.05 USD
100199-514 100206-964
Titanium(IV) isopropoxide ≥98%
Titanium (IV) isopropoxide

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formula: C₁₂H₂₈O₄Ti
MW: 284.23 g/mol
Boiling Pt: 58 °C (1 mmHg)
Melting Pt: 20 °C
Density: 0.9550
Flash Pt: 81 °F
MDL Number: MFCD00008871
CAS Number: 546-68-9
EINECS: 208-909-6
UN: 1993
ADR: 3,III
Merck Index: 12,09614

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR