Calcium fluoride ≥99.9% (Ca basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-2017-250G 93-2017-1000G
100210-240EA 243.45 USD
100210-240 100202-440
Calcium fluoride ≥99.9% (Ca basis)
Calcium fluoride
Formula: CaF₂
Boiling Pt: ~2500 °C
Melting Pt: 1360 °C
Density: 3.180
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR