Potassium ≥99.95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Prescored ampoule.
93-1990-5G
93-1990-1G
93-1990-25G
100203-840EA
152.86
USD
100203-840
100202-532
100205-866
Potassium ≥99.95%
Potassium
Formula:
K MW: 39.1 g/mol Boiling Pt: 760 °C Melting Pt: 63 °C Density: 0.86 Storage Temperature: Ambient |
MDL Number:
MFCD00133776 CAS Number: 7440-09-7 EINECS: 231-119-8 UN: 2257 ADR: 4.3,I Merck Index: 12,07763 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...