Potassium chloride ≥99% (99.99+%-K), PURATREM, Optical Grade

Supplier: STREM CHEMICALS INC MS
Warning

93-1977-50G 93-1977-10G
100198-326EA 225.05 USD
100198-326 100208-516
Potassium chloride ≥99% (99.99+%-K), PURATREM, Optical Grade
Potassium chloride
Formula: KCl
Boiling Pt: 1500 °C (subl.)
Melting Pt: 776 °C
Density: 1.984
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR