Potassium bromide 99,999 K basis, PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

93-1962-10G 93-1962-50G
100207-792EA 205.23 USD
100207-792 100206-014
Potassium bromide 99,999 K basis, PURATREM
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.75
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR