Potassium iodide ≥99.99% (K basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dipotassium diiodide, Kali iodide

93-1930-50G 93-1930-10G
100201-948EA 407.63 USD
100201-948 100209-032
Potassium iodide ≥99.99% (K basis), PURATREM
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Density: 3.13
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR