Potassium hexafluorophosphate ≥99.5%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1926-100G
93-1926-500G
100196-524EA
66.52
USD
100196-524
100196-526
Potassium hexafluorophosphate ≥99.5%
Potassium hexafluorophosphate
Formula:
F₆KP Boiling Pt: Dec. Melting Pt: ∼ 575 °C Density: 2.55 |
MDL Number:
MFCD00011412 CAS Number: 17084-13-8 EINECS: 241-143-0 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...