Potassium cyanide ≥96% ACS

Supplier: STREM CHEMICALS INC MS
Danger

93-1916-250G 93-1916-1000G
100204-204EA 169.85 USD
100204-204 100207-276
Potassium cyanide ≥96% ACS
Potassium cyanide
Formula: KCN
Boiling Pt: 1625 °C (1013 hPa)
Melting Pt: 634.5 °C
Density: 1.52
Storage Temperature: Ambient
MDL Number: MFCD00011397
CAS Number: 151-50-8
EINECS: 205-792-3
UN: 1680
ADR: 6.1,I
Merck Index: 13,07709

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR