Potassium chloride ≥99% ACS

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Potassiumchloride

93-1914-500G 93-1914-2000G
100200-060EA 69.35 USD
100200-060 100198-322
Potassium chloride ≥99% ACS
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420 (subl.)
Melting Pt: 773
Density: 1.984
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR