Dimethyl methylphosphonate ≥97%

Supplier: STREM CHEMICALS INC MS

93-1523-2000G 93-1523-100G 93-1523-500G
100196-516EA 394.89 USD
100196-516 100205-366 100203-394
Dimethyl methylphosphonate ≥97%
Dimethyl methylphosphonate
Formula: C₃H₉O₃P
MW: 124.08 g/mol
Boiling Pt: 181
Density: 1.160
Flash Pt: 156 °F
MDL Number: MFCD00008349
CAS Number: 756-79-6
EINECS: 212-052-3
Merck Index: 13,03425

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR