Chlorotrimethylsilane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: TMCS, TMSC, Trimethylchlorosilane

93-1464-100G 93-1464-500G
100200-054EA 67.94 USD
100200-054 100203-390
Chlorotrimethylsilane ≥97%
Chlorotrimethylsilane
Formula: C₃H₉ClSi
Boiling Pt: 57.3 °C
Melting Pt: –40 °C
Density: 0.8536 (27°)
Flash Pt: –18 °F
Storage Temperature: Ambient
MDL Number: MFCD00000502
CAS Number: 75-77-4
EINECS: 200-900-5
UN: 1298
ADR: 3,II
REACH: 01-2119457596-25

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR