Tetraethyl orthosilicate (TEOS) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tetraethoxysilane

93-1454-2000G 93-1454-500G
100205-860EA 345.35 USD
100205-860 100205-346
Tetraethyl orthosilicate (TEOS) ≥98%
Tetraethyl orthosilicate (TEOS)

Metal Alkoxides, Volatile Precursors for CVD

Formula: Si(OC₂H₅)₄
Boiling Pt: 165.8 °C
Melting Pt: –77 °C
Density: 0.934
Flash Pt: 116 °F
Storage Temperature: Ambient
MDL Number: MFCD00009062
CAS Number: 78-10-4
EINECS: 201-083-8
UN: 1292
ADR: 3,III
Merck Index: 13,03882

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR