Aluminium chloride, anhydrous ≥99%, Reagent Grade
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Aluminum chloride
93-1387-250G
93-1387-1000G
100205-322EA
121.72
USD
100205-322
100202-528
Aluminium chloride, anhydrous ≥99%, Reagent Grade
Aluminium chloride
Formula:
AlCl₃ MW: 133.34 g/mol Boiling Pt: 180 (subl.) Melting Pt: 190 °C Density: 2.44 Storage Temperature: Ambient |
MDL Number:
MFCD00003422 CAS Number: 7446-70-0 EINECS: 231-208-1 UN: 1726 ADR: 8 II Merck Index: 13,00335 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...