Aluminium chloride hexahydrate ≥99%, Reagent Grade

Supplier: STREM CHEMICALS INC MS
Danger

93-1309-250G 93-1309-1000G
100203-372EA 110.4 USD
100203-372 100203-838
Aluminium chloride hexahydrate ≥99%, Reagent Grade
Aluminium chloride hexahydrate
Formula: AlCl₃·6H₂O
Melting Pt: 100 °C (dec.)
Density: 2.398
Storage Temperature: Ambient
MDL Number: MFCD00149134
CAS Number: 7784-13-6
EINECS: 231-208-1
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR