Magnesium ≥99.8%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1289-1000G
93-1289-250G
100200-314EA
352.43
USD
100200-314
100200-510
Magnesium ≥99.8%, powder
Magnesium
Formula:
Mg MW: 24.3 g/mol Boiling Pt: 1107 Melting Pt: 651 Density: 1.74 Storage Temperature: Ambient |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1418 ADR: 4.3 II Merck Index: 13,05675 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...