Magnesium oxide ≥99.95% (Mg basis) -325 mesh

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Magnesia usta

93-1241-100G 93-1241-25G
100208-094EA 462.83 USD
100208-094 100208-096
Magnesium oxide ≥99.95% (Mg basis) -325 mesh
Magnesium oxide
Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600
Melting Pt: 2800
Density: 3.58
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR