Sodium tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Sodium fluoroborate

93-1189-500G 93-1189-2000G
100196-472EA 108.98 USD
100196-472 100208-090
Sodium tetrafluoroborate ≥98%
Sodium tetrafluoroborate
Formula: BF₄Na
MW: 109.79 g/mol
Melting Pt: 384 (dec.)
Density: 2.47
Storage Temperature: Ambient
MDL Number: MFCD00003515
CAS Number: 13755-29-8
EINECS: 237-340-6
UN: 1759
ADR: 8 II
Merck Index: 12,08761

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR