di-Sodium tetraborate, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Borax, Sodium tetraborate

93-1188-500G 93-1188-2000G
100205-288EA 43.88 USD
100205-288 100209-004
di-Sodium tetraborate, anhydrous ≥99%
di-Sodium tetraborate
Formula: Na₂B₄O₇
Boiling Pt: 1575 °C (dec.)
Melting Pt: 741 °C
Density: 2.367
Storage Temperature: Ambient
MDL Number: MFCD00081185
CAS Number: 1330-43-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 12,08733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR