Sodium dihydrogen phosphate monohydrate ≥99.9% (Na basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
93-1174-1000G
93-1174-250G
100198-294EA
225.05
USD
100198-294
100201-866
Sodium dihydrogen phosphate monohydrate ≥99.9% (Na basis)
Sodium dihydrogen phosphate monohydrate
Formula:
NaH₂PO₄·H₂O Melting Pt: 100 °C Density: 2.04 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149208 CAS Number: 10049-21-5 EINECS: 231-449-2 Merck Index: 13,08734 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...