Sodium hydride 60% in oil

Supplier: STREM CHEMICALS INC MS
Danger

93-1151-1000G 93-1151-250G
100206-906EA 368 USD
100206-906 100196-456
Sodium hydride 60% in oil
Sodium hydride
Formula: HNa
Storage Temperature: Ambient
MDL Number: MFCD00003471
CAS Number: 7646-69-7
EINECS: 231-587-3
UN: 1427
ADR: 4.3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR